Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Disodium metasilicate pentahydrate, Technical Grade

Supplier: BeanTown Chemical

225820-1KG
BT225820-1KGEA 49.71 USD InStock
BT225820-1KG
Disodium metasilicate pentahydrate, Technical Grade
di-Sodium metasilicate pentahydrate
  • RTECS: VV9287500
  • -18 Mesh Granular


Formula: Na₂SiO₃·5H₂O
MW: 212.14 g/mol
Melting Pt: 72 °C
MDL Number: MFCD00149176
CAS Number: 10213-79-3
EINECS: 229-912-9
UN: 3253
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty