Would you like to log in again?
Whatman™ Grade 6 Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
1006-240 1006-150 1006-090 1006-070 1006-042 1006-185 1006-125 1006-110 1006-125 1006-042 1006-185 1006-070 1006-110 1006-240 1006-150 1006-090
10147-992PK 79.09 USD InStock
10147-992 10147-990 10147-994 CA10147-992 CA11008-570 CA11012-566 CA11008-568 CA11008-516 CA28462-170 10147-902 28462-170 10147-988 CA10147-904 CA10147-902 10147-904 10035-236
Whatman™ Grade 6 Qualitative Filter Papers, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
A standard grade filter for boiler water analysis.
  • Twice as fast as Grade 5 with similar fine particle retention
  • Often specified for boiler water analysis applications
These cellulose filters are used in qualitative analytical techniques to determine and identify materials. Prepleated qualitative filters are also available, which give improved flow rate and increased loading capacity compared to equivalent flat filters.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty