Would you like to log in again?
Alkaline Phosphatase
Supplier: Cytiva
E70092Z E70092X E70092Y E70092Z E70092X E70092Y
95040-324EA 229.84 USD InStock
95040-324 CA95040-324L CA95040-320L 95040-322 95040-320 CA95040-322L
Alkaline Phosphatase
Enzymes
A high specific activity, heat-labile alkaline phosphatase. Completely and irreversibly inactivated in Tris buffers at pH 8,0 to 8,5 by heating for 15 minutes at 65 °C. No further treatment is necessary.
  • Bacterial alkaline phosphatase recombinantly expressed in E coli
  • Active in PCR and restriction endonuclease buffers
  • Inactivated by heating at 75°C for 5 minutes
  • Easily soluble in cold water and hot water
  • Sourced from arctic shrimp (Pandalus borealis)
Use together with illustra Exonuclease I for cleaning up PCR reactions prior to downstream application such as DNA sequencing. Dephosphorylation of a restriction enzyme digested plasmid (5-20 pmol of 5'-ends, 0.1-0.5 units/pmol 5'-ends). Reduces re-ligation to < 0.5% compared to the untreated control.
One unit is the amount of the enzyme required to dephosphorylate 1 µg of linearized pUC57 DNA termini in 10 minutes at 37°C. The enzyme is purified to apparent homogeneity and is free of all contaminating endonucleases, exonucleases and ribonucleases
Unit Definition: One unit is the amount of the enzyme required to dephosphorylate 1 Â µg of linearized pUC57 DNA termini in 10 minutes at 37 °C.
Ordering information: Includes 10x reaction buffer (200 mM Tris-HCl, pH 8.0, 100 mM MgCl2) and dilution buffer (50 mM Tris-HCl, pH 8.0)
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty