Would you like to log in again?
Connectors, Luer, 1/16", Cytiva
Supplier: Cytiva
18-1112-51 18-1112-51 18-1112-51
10500-512EA 110.13 USD InStock
10500-512 10500-513 CA10500-512
Connectors, Luer, 1/16", Cytiva
Fittings and Connectors
Designed to provide connection of syringe to top of column
When ordered separately the connectors are black.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty