Would you like to log in again?
Whatman™ Seed Testing Papers, Grade 181, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
2181-090 2181-090
10035-738PK 46.1 USD InStock
CA11013-698 10035-738
Whatman™ Seed Testing Papers, Grade 181, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
Grade 181 support filter for seed tests using copenhagen tank system.
  • Extremely high purity
  • Reliable and reproducible results
The papers are made from pure cellulose without any additives and do not contain any substances that could influence the growth of the seeds. The constant water absorption of the papers ensures the continuous provision of the required amount of water.
The better contrast of the color seed testing papers makes evaluation easier, particularly for seeds with fine white rootlets or under artificial light. This makes work easier, improves the results, and saves time. The dyes that we use have been thoroughly investigated and have no influence on the growth of the seeds.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty