Would you like to log in again?
Whatman™ Grade 91 Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
1091-240 1091-240 1091-930 1091-185 1091-125 1091-930 1091-125 1091-185 1091-190 1091-165 1091-110 1091-150 1091-165 1091-935 1091-190 1091-150 1091-110 1091-935
10035-708PK 444.49 USD InStock
CA11013-316 10035-708 CA11013-304 CA11013-314 10035-704 CA11013-310 CA11013-288 10035-706 CA11013-364 CA11013-286 CA11013-292 10035-240 10035-710 10035-238 10035-700 10035-712 10035-702 CA11013-308
Whatman™ Grade 91 Qualitative Filter Papers, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
A general purpose creped cellulose filter for less critical routine analysis. Widely used to assay sucrose in cane sugar and within pharmaceutical laboratories for routine filtration.
These extremely strong filter papers have a high wet strength due to the addition of a small quantity of chemically stable resin. Normal qualitative applications will not introduce any significant impurities into the filtrate. The resins do, however, contain nitrogen so these grades should not be used in Kjeldahl estimations, etc.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty