Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whatman™ Grade GF 9 Glass Microfiber Filters with Binder, Whatman products (Cytiva)

Supplier: Whatman products (Cytiva)
Whatman™
10370205 10370206 10370205 10370206 10370202 10370202
10034-844PK 275.5 USD InStock
CA11013-854 CA11013-456 10034-844 10034-842 CA11013-418 10034-840
Whatman™ Grade GF 9 Glass Microfiber Filters with Binder, Whatman products (Cytiva)
Filters Glass Fiber Filters
Whatman Grade GF 9 glass microfiber filters from Cytiva's business are strengthened with an inorganic binder for environmental analysis of coarse, heavy samples.

  • Retention of coarse particles (~ 3 µm)
  • Strengthened with inorganic binder
  • Can withstand temperatures up to 500 °C
  • Medium filtration speed
  • Can be used as a pre-filter for dense samples

Cytiva provides a number of binder-strengthened filters for use in varied environmental analyzes.

Grade GF 9 borosilicate glass filters can be used in a range of environmental analysis applications. The added strength of the inorganic binder makes these filters appropriate for use in retention of samples with heavy particles.

Other applications include:
- Determination of Polychlorinated biphenyl (PCB), Dichlorodiphenyldichloroethylene (DDE), Dichlorodiphenyltrichloroethane (DDT), furans, and dioxins in air
- Pollution measurements in industrial, urban, and populated areas
- Dust measurements in the workplace
- Determination of dust fraction in technical gases; Testing the effectiveness of dust-collecting

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty