Would you like to log in again?
XK and K 9 Series O-Rings, Cytiva
Supplier: Cytiva
19068501
101724-964EA 172.28 USD InStock
101724-964
XK and K 9 Series O-Rings, Cytiva
Seals Sealing Rings
Can be used in aqueous solutions and nearly all organic solvents commonly used in liquid chromatography of macromolecules with the following few exceptions - chlorinated hydrocarbons, acetone and other ketones, aliphatic esters and phenol
Solutions containing more than 10% NaOH, 10% HCl, other strong mineral acids or 5% acetic acid should not be used.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty