Would you like to log in again?
Tricorn Filter Kits
Supplier: Cytiva
Tricorn™
29053612 11001253 29053612 29053586 29053586
101724-576EA 226.2 USD InStock
101724-576 76488-166 76488-168 CA76488-166 CA76488-168
Tricorn Filter Kits
Filters Filter Discs
Filter kits for Tricorn 5 and 10 columns are available in three porosities.
Choose the standard 7 µm porosity for most applications. Use the medium 18 µm porosity for the purification of polyclonal IgG antibodies or mAbs in combination with resins such as MabSelect SuRe pcc or MabSelect PrismA. Use the coarse filter (>25 µm) to reduce the risk of clogging your capture resin when working with large volumes or repeated loadings of clarified samples.
Delivery information: All filter kits include top and bottom filters and ethylene propylene diene monomer (EPDM) O-rings, five of each.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty