Would you like to log in again?
HiScale Adapter, Complete, Cytiva
Supplier: Cytiva
HiScale™
28966383
101725-280EA 741.54 USD InStock
101725-280
HiScale Adapter, Complete, Cytiva
Chromatography Supplies Chromatography Spare Parts
Can be used in aqueous solutions and nearly all organic solvents commonly used in liquid chromatography of macromolecules with the following few exceptions - Chlorinated hydrocarbons, acetone and other ketones, aliphatic esters and phenol.
Solutions containing more than 10% NaOH, 10% HCl, other strong mineral acids or 5% acetic acid should not be used. Includes 1 x net (20 µm), 1 x support screen, 1 x distributor, 1 x o-ring 10.3 x 2.4 mm (FPM), 1 x Ferrule 1/16", 30 cm PEEK tubing i.d. 1.0 mm (1/16"), 1 x inner shaft, 1 x adapter shaft, 1 x antirotator, 1 x end cap, 1 x end knob, 50 mm silicone tubing i.d. 2.1 mm, o.d. 4.2 mm, 1 x domed nut, M6, 1 x fingertight connector, 1 x union f/f and 1 x fingertight stop plug.
Maximum temperature: 4 to 40 °C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty