Would you like to log in again?
Screw Lid Kit, Cytiva
Supplier: Cytiva
11-0004-10 11-0004-10
10498-012EA 60.01 USD InStock
10498-012 CA10498-012
Screw Lid Kit, Cytiva
Closures Bottle Closures
The Screw Lid Kit is a bottle-top attachment for buffer bottles used with AKTA systems.
The polypropylene lid is sized for GL45 glass bottles with a membrane that allows tubing to be passed through. The kit contains one screw lid and one cap membrane. The membrane should not be used when working with organic solvents.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty