Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Superloop Complete, 150 ml, Cytiva

Supplier: Cytiva

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

18-1023-85 18-1023-85
10498-014EA 2125.63 USD InStock
CA10498-014 10498-014
Superloop Complete, 150 ml, Cytiva
Chromatography Supplies Chromatography Spare Parts
The glass tube has a graduated scale of 5 ml increments and seals against the inner end pieces

A springloaded valve made from titanium is located in the center of the movable seal. When Superloop is in loading position, the valve is closed and remains closed during sample loading. When the sample is injected onto the column the seal moves downwards. When the bottom position is reached, the pressure increase causes the valve to open and the buffer to pass through the valve. Superloop is for use with aqueous solutions and alcohols.

Ordering information: Tubing and connectors have to be ordered separately.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty