Would you like to log in again?
Thomson SINGLE StEP Standard Filter Vials, Restek
Supplier: Restek
25894 25897
10118-322PK 457.97 USD InStock
10118-322 10118-326
Thomson SINGLE StEP Standard Filter Vials, Restek
Filters Syringeless Filters
Recommended for samples containing less than 10% solid particulates, these standard filter vials are constructed using PTFE or PES for long-lasting durability.
  • Minimize sample loss by eliminating multiple transfers
  • Color-coded caps allow easy identification of 0.2 or 0.45 μm membranes in PVDF, PTFE, PES, or nylon
  • Pre-slit PTFE/silicone caps help eliminate broken autosampler needles and cored septa
  • Rugged polypropylene vial houses insert with 450 μl loading capacity and low dead volume (120 μl)
  • Fit most standard 12×32 mm autosamplers, including UHPLC instruments
Easy-to-use vials offer fast sample filtration and require only a squeeze of your fingers.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty