Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4-Difluorophenylmagnesium bromide 0.5 M in 2-MeTHF (2-methyltetrahydrofuran)

Supplier: Thermo Scientific Chemicals
Danger

H54197-AE H54197-AE
AAH54197-AEEA 274.99 USD InStock
CAAAH54197-AE AAH54197-AE
3,4-Difluorophenylmagnesium bromide 0.5 M in 2-MeTHF (2-methyltetrahydrofuran)
3,4-Difluorophenylmagnesium bromide


Formula: C₆H₃BrF₂Mg
MW: 217.296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00672004
CAS Number: 90897-92-0
UN: 2924
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty