Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals
Synonyms: 4-Amino-2-fluoro-5-methoxyphenylboronic acid pinacol ester5-fluoro-2-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

H59944-03 H59944-03 H59944-MD H59944-MD
AAH59944-03EA 517.99 USD InStock
AAH59944-03 CAAAH59944-03 AAH59944-MD CAAAH59944-MD
4-Amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester 96%
4-Amino-2-fluoro-5-methoxybenzeneboronic acid pinacol ester


Formula: C₁₃H₁₉BFNO₃
MW: 267.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22419265
CAS Number: 1326283-60-6
Order Now
Specification Test Results
Assay Minimum 96

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty