Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

H-D-Asp-OMe 98%

Supplier: Thermo Scientific Chemicals
Synonyms: D-Aspartic acid 1-methyl ester(R)-3-Amino-4-methoxy-4-oxobutanoic acid

H63658-06 H63658-03 H63658-14
AAH63658-06EA 83.49 USD InStock
AAH63658-06 AAH63658-03 AAH63658-14
H-D-Asp-OMe 98%
H-D-Asp-OMe


Formula: C₅H₉NO₄
MW: 147.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03424227
CAS Number: 65414-78-0
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty