Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Serine benzyl ester hydrochloride 95%

Supplier: Thermo Scientific Chemicals
Synonyms: H-D-Ser-OBzl.HClH-D-Ser-OBzl·HCl

H63678-06 H63678-03
AAH63678-03EA 94.68 USD InStock
AAH63678-03 AAH63678-06
D-Serine benzyl ester hydrochloride 95%
D-Serine benzyl ester hydrochloride


Formula: C₁₀H₁₄ClNO₃
MW: 231.676 g/mol
Melting Pt: 169…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00191017
CAS Number: 151651-44-4
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty