Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trichloroacetic acid (TCA) 10% (w/v) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Aceto-CaustinTCATrichloroethanoic acid

J62355-AP J62355-AP
AAJ62355-APEA 141.6 USD InStock
CAAAJ62355-AP AAJ62355-AP
Trichloroacetic acid (TCA) 10% (w/v) in aqueous solution
Trichloroacetic acid (TCA)
Protein precipitation reagent


Formula: C₂HCl₃O₂
MW: 163.39 g/mol
Melting Pt: 54…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
EINECS: 200-927-2
UN: 2564
ADR: "8",III
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/52/727452.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty