Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Hydroxysaclofen

Supplier: Thermo Scientific Chemicals
Danger

J63588-MA J63588-MA
AAJ63588-MAEA 512.35 USD InStock
CAAAJ63588-MA AAJ63588-MA
2-Hydroxysaclofen
2-Hydroxysaclofen
Selective antagonist at GABA-B receptors

Solid


Formula: C₉H₁₂ClNO₄S
MW: 265.708 g/mol
Melting Pt: 268…271 °C
Storage Temperature: Ambient
MDL Number: MFCD00069282
CAS Number: 117354-64-0
UN: 1759
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty