Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(-)-2-Amino-5-phosphonopentanoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: D-AP5D-2-Amino-5-phosphonovaleric acid

J64887-MA J64887-MA
AAJ64887-MAEA 329.68 USD InStock
AAJ64887-MA CAAAJ64887-MA
(R)-(-)-2-Amino-5-phosphonopentanoic acid 99%
(R)-(-)-2-Amino-5-phosphonopentanoic acid
A competitive NMDA antagonist


Formula: C₅H₁₂NO₅P²
MW: 197.127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078839
CAS Number: 79055-68-8
Order Now
Specification Test Results
Assay Minimum 99

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty