Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chrysophenine

Supplier: Thermo Scientific Chemicals
Synonyms: Chrysophenine sodium salt

J64999-18 J64999-30 J64999-18 J64999-22
10814-360EA 283.8 USD InStock
10814-360 AAJ64999-18 10814-358 CAAAJ64999-18
Chrysophenine
Chrysophenine
A diazo stilbene dye

Powder


Formula: C₃₀H₂₈N₄O₈S₂
MW: 680.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007488
CAS Number: 2870-32-8
EINECS: 220-698-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty