Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Casein Sodium Salt

Supplier: Thermo Scientific
J65590-36 J65590-36 J65590-A1 J65590-A1
AAJ65590-A1EA 73.39 USD InStock
CAAAJ65590-A1 CAAAJ65590-36 AAJ65590-A1 AAJ65590-36
Casein Sodium Salt
Proteins and Peptides
Powder

CAS No.: 9005-46-3
Physical Form: Powder
Appearance: Off-white
MDL No.: MFCD00130736

Applications: Casein is frequently used as a blocking agent in immunochemistry experiments. It is also used in the recovery of enzyme activity from SDS extracted samples, and as a substrate for protease and kinase assays.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty