Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gold 99% supplied in 0.1 mM PBS, nanoparticles 150 nm reactant-free

Supplier: Thermo Scientific Chemicals

J67040-AC J67040-AE
AAJ67040-AEEA 409.86 USD InStock
AAJ67040-AE AAJ67040-AC
Gold 99% supplied in 0.1 mM PBS, nanoparticles 150 nm reactant-free
Gold
Ideal for sensitive applications requiring a minimum amount of contaminating reactants. Well suited for protein conjugate development by passive adsorption to the gold nanoparticle surface. Conjugates can be used in blotting, electron microscopy and immunohistochemistry applications

Liquid


Formula: Au
MW: 196.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
Order Now
Specification Test Results
Assay Minimum 99

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty