Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gold supplied in purified water biotin-functionalized, PEGylated, nanoparticles 100 nm OD50 (Abs. 572 nm)

Supplier: Thermo Scientific Chemicals

J67145-AMI J67145-XF
AAJ67145-AMIEA 339.74 USD InStock
AAJ67145-AMI AAJ67145-XF
Gold supplied in purified water biotin-functionalized, PEGylated, nanoparticles 100 nm OD50 (Abs. 572 nm)
Gold


Formula: Au
MW: 196.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty