Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boric acid ≥99.5% for electrophoresis

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Boracic acidBoron hydroxideBoron trihydroxideOrthoboric acid

J67202-A7 J67202-36 J67202-A1
AAJ67202-36EA 55.28 USD InStock
AAJ67202-36 AAJ67202-A1 AAJ67202-A7
Boric acid ≥99.5% for electrophoresis
ortho-Boric acid
Powder


Formula: B(OH)₃
MW: 61.83 g/mol
Melting Pt: ca 185 °C (dec.)
Density: 1.435
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/54/16640554.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty