Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

NOB 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 4-Nitro-3-(octanoyloxy)benzoic acid

J67360-MF
AAJ67360-MFEA 188.7 USD InStock
AAJ67360-MF
NOB 98%
NOB
A phospholipase substrate

Solid


Formula: C₁₅H₁₉NO₆
Storage Temperature: Ambient
MDL Number: MFCD00132973
CAS Number: 55894-52-5
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty