Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boric acid ≥99.5% for molecular biology

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Boracic acidBoron hydroxideBoron trihydroxideOrthoboric acid

J67419-A1 J67419-A7 J67419-36
AAJ67419-36EA 67.86 USD InStock
AAJ67419-36 AAJ67419-A1 AAJ67419-A7
Boric acid ≥99.5% for molecular biology
ortho-Boric acid
Powder


Formula: B(OH)₃
MW: 61.83 g/mol
Melting Pt: ca 185 °C (dec.)
Density: 1.435
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/54/16640554.jpg
Order Now
Specification Test Results
Assay Minimum 99.5

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty