Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gold 95% supplied in 0.1 mM PBS, nanoparticles 60 nm OD1 (Abs. 540 nm)

Supplier: Thermo Scientific Chemicals

J67438-AC J67438-AE
AAJ67438-AEEA 296.09 USD InStock
AAJ67438-AE AAJ67438-AC
Gold 95% supplied in 0.1 mM PBS, nanoparticles 60 nm OD1 (Abs. 540 nm)
Gold
For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM

Liquid


Formula: Au
MW: 196.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty