Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Arachidonylglycine

Supplier: Thermo Scientific Chemicals

J67492-LB0
AAJ67492-LB0EA 152.16 USD InStock
AAJ67492-LB0
N-Arachidonylglycine
N-Arachidonylglycine
Waxy solid


Formula: C₂₂H₃₅NO₃
MW: 361.526 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03791313
CAS Number: 179113-91-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty