Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver conductive coating

Supplier: Thermo Scientific Chemicals
Danger

44937-10 044937-10
AA44937-10EA 195.99 USD InStock
AA44937-10 CAAA44937-10
Silver conductive coating
Silver (gold coated)
Sheet resistance 0.015 ohms/square in./mil (25um).


Formula: Ag
Boiling Pt: 56 °C
Density: 1.62 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
UN: 1133
ADR: 3,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty