Would you like to log in again?
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), gauze woven from wire 80 mesh Ø 0.14 mm (0.0055 in) Type 316
Supplier: Thermo Scientific Chemicals
Danger
046574-HB 046574-FL 46574-HB 46574-GT 46574-FL 046574-GT
AA46574-FLEA 26.79 USD InStock
AA46574-FL CAAA46574-FL AA46574-HB CAAA46574-HB AA46574-GT CAAA46574-GT
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), gauze woven from wire 80 mesh Ø 0.14 mm (0.0055 in) Type 316
Stainless steel
Wire Cloth
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty