Would you like to log in again?
HotSeal™ Sealing Film, Diversified Biotech
Supplier: Diversified Biotech
HOTS-1000
10141-846PK 113.15 USD InStock
10141-846
HotSeal™ Sealing Film, Diversified Biotech
Seals Microplate Seals
Prevents well-to-well contamination, evaporation and spillage. Ideal for PCR applications.
  • RNase and DNase free
  • Strong 2.3 mil thick polyester film
  • Non-cytotoxic and sterile
  • Functional from -40°C to +125°C
  • Two scored end tabs for easy positioning
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty