Would you like to log in again?
Solvent Resistant Tags, Diversified Biotech
Supplier: Diversified Biotech
Tough-Tags®
RVTH-1000 RVTH-3000 RVTH-2000 RVHD-2000 RVHD-1000
10141-774RL 57.46 USD InStock
10141-774 10141-782 10141-780 10141-778 10141-776
Solvent Resistant Tags, Diversified Biotech
Labels
Solvent resistant labels that cleanly remove from surfaces.
  • Resistant to most laboratory solvents and have shown short-term resistance to Acetone and Xylene
  • When direct thermal printed or mark with a solvent resistant pen, labels exhibit exceptional durability
  • Available on rolls for hand writing or for direct thermal printing using the Dymo® LabelWriter
  • Temperature resistant from –80 to 93 °C
Great for beakers, bottles, flasks, glass vials, tubes, tube racks and pipettes and easily remove from surfaces without leaving any adhesive.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty