Would you like to log in again?
Thermal Transfer Cryogenic Tags, Diversified Biotech
Supplier: Diversified Biotech
Cryo-Tags®
CRTH-3150 CRTH-3128 CRTH-3225 CRTH-3094 CRTH-3081 CRTH-4000 CRTH-3155 CRTH-2300
10141-708RL 259.38 USD InStock
10141-708 10141-716 10141-706 10141-714 10141-704 10141-712 10141-702 10141-700
Thermal Transfer Cryogenic Tags, Diversified Biotech
Labels
Thermal transfer Cryo-Tags® withstand long term cryogenic and freezer storage.
Labels are chemically inert with solvent resistant adhesive that adheres to croygenic vials, microtubes, test tubes, and metal racks. Cryo-Tags® withstand flashfreezing in liquid nitrogen, and dry heat in the range of –190 to 80 °C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty