Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TRKB Rabbit Polyclonal Antibody

Supplier: BioVision
AB18987-100
10004-328EA 937.37 USD InStock
10004-328
Anti-TRKB Rabbit Polyclonal Antibody
Antibodies
Due to the various splice variants, the trk proteins exist as variably glycosylated entities with the major forms having molecular weights of 140-145 kDa, 110 kDa and the unglycosylated form of 80 kDa. trkB contains 33.3% carbohydrate by weight representing modification on 10 of 12 N-glycosylation sites. The primary ligand for trkB is BDNF which induces the phosphorylation of the protein and subsequent binding of PLC-gamma via SH2 domains. TrkB may function to modulate neuronal responses to the neurotrophins acting throμgh trkB such as BDNF.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Iimmunofluorescence: 5-10 µg/ml

Type: Primary
Antigen: TRKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Chicken

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty