Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CASP9 Rabbit Polyclonal Antibody

Supplier: BioVision
AB286147-100UG
10003-220EA 896.32 USD InStock
10003-220
Anti-CASP9 Rabbit Polyclonal Antibody
Antibodies
Caspase-9 is one of the most important caspases among the caspase family members. Caspase-9 is synthesized as inactive pro-enzyme that is processed in cells undergoing apoptosis. The processed caspase-9 consists of large (32-35 kD) and small (10 kD) subunits which associate to form an active enzyme. Activated caspase-9 in turn cleaves and activates caspase-3, one of the proteases responsible for the proteolytic cleavage of many key proteins in apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CASP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Hamster, Bovine, Rabbit, Pig

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty