Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CASP11 Rabbit Polyclonal Antibody

Supplier: BioVision
AB18741-100
10005-466EA 360.69 USD InStock
10005-466
Anti-CASP11 Rabbit Polyclonal Antibody
Antibodies
Caspase family of cysteine proteases has been shown to play a key role in apoptosis. Caspase-11 is also synthesized as inactive pro-enzyme that are processed in cells undergoing apoptosis or inflammation. The processed form of caspase-11 consists of large and small subunits which associate to form an active enzyme.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: CASP11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty