Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AKT1 Rabbit Polyclonal Antibody

Supplier: BioVision
AB18785-100
10003-672EA 373.8 USD InStock
10003-672
Anti-AKT1 Rabbit Polyclonal Antibody
Antibodies
Akt, also referred to as PKB and RAC, is the cellular homologue of the viral oncogene v-Akt. This protein kinase is activated by insulin and various growth factors and functions in a pathway involving PI3 kinase as activation is sensitive to wortmanin. Recent evidence sμggests that Akt functions to promote cell survival by actively inhibiting apoptosis.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; in Immunoprecipitation: 10-40 µg/ml; and Immunohistochemistry: 10-40 µg/ml

Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty