Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFA Rabbit Polyclonal Antibody

Supplier: BioVision
AB286149-100UG
10003-268EA 828.86 USD InStock
10003-268
Anti-TNFA Rabbit Polyclonal Antibody
Antibodies
Tumor Necrosis Factor-alpha (TNF-α) is a potent lymphoid factor that exerts cytotoxic effects on a wide range of tumor cells and certain other target cells. Rat TNF-α is a 17 kDa protein containing 157 amino acid residues.

Recommended Dilutions: Western Blot: 1-2 µg/ml; Immunohistochemistry: 10-20 µg/ml; Flow Cytometry: 10-20 µg/ml; Neutralization: 0.5 µg/ml

Type: Primary
Antigen: TNFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty