Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SGLT2 Rabbit Polyclonal Antibody

Supplier: BioVision
AB37296-100
10004-544EA 406.58 USD InStock
10004-544
Anti-SGLT2 Rabbit Polyclonal Antibody
Antibodies
Sodium glucose co transporters (SGLTs) are located in the brush-border membrane, but transported out of the cell across the basolateral membranes by a facilitated sμgar transporter (GLUT). At least three members of SGLTs (SGLT 1-3) have been cloned and characterized. SGLTs transport a methyl-D-glucoside (a-MDG), a non-metabolized model substrate, in Na-dependent manner. SGLT-1 does not discriminate a-MDG, glucose, and galactose. SGLT2/3 do not transport D-galactose efficiently.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: SGLT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty