Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Tubulin Rabbit Polyclonal Antibody

Supplier: BioVision
AB59680-100
10005-888EA 379.77 USD InStock
10005-888
Anti-Tubulin Rabbit Polyclonal Antibody
Antibodies
Microtubules are one type of cytoskeleton. Globular tubulin subunits are the building blocks of a microtubule and the α and β-tubulin heterodimer forms the tubulin subunits, which is present in all eukaryotic cells. Microtubules mediate many cell movements.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Tubulin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty