Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TFP1/HADHA Rabbit Polyclonal Antibody

Supplier: BioVision
AB54477-100
10004-670EA 393.47 USD InStock
10004-670
Anti-TFP1/HADHA Rabbit Polyclonal Antibody
Antibodies
Mitochondrial Trifunctional Protein (TFP) is a multienzyme complex of the beta-oxidation cycle. TFP deficiency is a clinically heterogeneous disorder with phenotypes of different severity. The spectrum of diseases range from severe neonatal/infantile cardiomyopathy and early death to mild chronic progressive sensorimotor poly-neuropathy with episodic rhabdomyolysis. Human TFP is an octomer composed of four alpha-subunits and four beta-subunits. Mutations in either subunits may result in general TFP deficiency with reduced activity of all enzymes.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TFP1/HADHA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty