Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Luciferase Rabbit Polyclonal Antibody

Supplier: BioVision
AB99944-200
10004-828EA 367.12 USD InStock
10004-828
Anti-Luciferase Rabbit Polyclonal Antibody
Antibodies
The Firefly luciferase is a genetically modified variant derived from the Luciferase of Diaphanes pectinealis endemic to Yunnan province, China. It has been modified to provide enhanced stability compared to the normal phenotype of Photinus pyralis as well as a broader and more physiologically relevant effective pH range. Luciferase has been used as a sensitive reporter for studying gene regulation and function. Firefly luciferase catalyzes the oxidative carboxylation of luciferin, a reaction with the highest efficiency of any known bioluminescence reaction.

Recommended Dilutions: Western Blot: 0.5-4 ?g/ml

Type: Primary
Antigen: Luciferase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Luciferase

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty