Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Relaxin-1 Rabbit Polyclonal Antibody

Supplier: BioVision
AB70803-100
10004-838EA 334.46 USD InStock
10004-838
Anti-Relaxin-1 Rabbit Polyclonal Antibody
Antibodies
Relaxin is a peptide hormone belonging to the insulin gene superfamily. It is made by the corpora lutea of the ovaries during pregnancy in many mammalian species. The secretion of the hormone into the blood stream just before parturition results in softening and lengthening of the pubic symphysis and a softening of the cervix, which facilitates the process of birth.
Relaxin-1 is proteolytically processed into a hetorodimer that has two peptide chains, A and B that are covalently linked by disulfide bonds.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: Relaxin-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty