Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Red Fluorescent Protein Mouse Monoclonal Antibody

Supplier: BioVision
AB50896-100
10005-190EA 393.47 USD InStock
10005-190
Anti-Red Fluorescent Protein Mouse Monoclonal Antibody
Antibodies
Since the molecular cloning of GFP cDNA and demonstration of GFP as a functional transgene, GFP has become a powerful tool with exciting applications in developmental, cell and molecular biology. RFP is the recent discovered protein that has the similar function and applications as GFP. Either GFP or RFP fluorescence is not species specific and can be expressed in bacteria, yeast, plant and mammalian cells. GFP or RFP can fuse with proteins of interest without interfering significantly with their assembly and function.

Recommended Dilutions: Western Blot: 1-4 ?g/ml; Iimmunofluorescence: 10-20 ?g/ml; Immunohistochemistry: 10-20 ?g/ml

Type: Primary
Antigen: Red Fluorescent Protein
Clonality: Monoclonal
Clone: N/A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: All Species

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty