Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GFP Rabbit Polyclonal Antibody

Supplier: BioVision
AB286193-100UG
10005-206EA 658.13 USD InStock
10005-206
Anti-GFP Rabbit Polyclonal Antibody
Antibodies
Green fluorescent protein (GFP) is a spontaneously fluorescent protein isolated from pacific jellyfish, Aequorea victoria. It transduces the blue chemiluminescence into green fluorescent light. Since the molecular cloning of GFP cDNA and demonstration of GFP as a functional transgene, GFP has become a powerful tool with exciting applications in developmental, cell and molecular biology. GFP fluorescence is not species specific and can be expressed in bacteria, yeast, plant and mammalian cells. GFP can fuse with proteins of interest without interfering significantly with their assembly and function.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; in Immunoprecipitation: 10-20 µg/ml

Type: Primary
Antigen: GFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: All Species

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty