Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SDF-1 beta Rabbit Polyclonal Antibody

Supplier: BioVision
AB18919-100
10007-760EA 413.14 USD InStock
10007-760
Anti-SDF-1 beta Rabbit Polyclonal Antibody
Antibodies
SDF-1β (stromal cell-derived factor-β), also known as pre-B-cell growth-stimulating factor (PBSF), is a 74-amino acid CXC chemokine originally cloned from a bone marrow stromal cell line. Targeted deletion of SDF-1 gene resulted in defects of B-cell lymphopoiesis and bone marrow myelopoiesis. SDF-1 has been shown to be chemotactic for lymphocytes. In addition, SDF-1 was recently reported to be a ligand for CXCR4 (LESTR/fusin), a co-receptor for HIV-1 entry into T cells. SDF-1 binding to CXCR4 inhibits HIV-1 entry.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml; Immunoprecipitation: 4-8 µg/ml; Immunohistochemistry: 10-20 µg/ml

Type: Primary
Antigen: SDF-1 beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty