Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TCRB Rabbit Polyclonal Antibody

Supplier: BioVision
AB18862-100
10007-892EA 334.46 USD InStock
10007-892
Anti-TCRB Rabbit Polyclonal Antibody
Antibodies
TCR (T cell antigen receptor) recognizes foreign antigens and translates such recognition events into intracellular signals that elicit a change in the cell from a dormant to an activated state. TCR is a heterodimer composed of either α and β or γ and δ chains. The vast majority of circulating T cells (95%) express the α/β heterodimer while roμghly 2-5% express the γ/δ heterodimer. CD3 chains and the CD4 or CD8 coreceptors are also required for efficient signal transduction throμgh the TCR.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: TCRB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty