Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VENTX Rabbit Polyclonal Antibody

Supplier: BioVision
AB124523-100UG
10003-752EA 796.73 USD InStock
10003-752
Anti-VENTX Rabbit Polyclonal Antibody
Antibodies
VENTX is a member of the Vent family of homeodomain proteins. VENTX may function as a transcriptional repressor and involved in mesodermal patterning and hemapoietic stem cell maintenance.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: VENTX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Xenopus

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty