Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HDAC10 Rabbit Polyclonal Antibody

Supplier: BioVision
AB18971-100
10004-384EA 347.56 USD InStock
10004-384
Anti-HDAC10 Rabbit Polyclonal Antibody
Antibodies
HDAC family are divided into two classes, I and II. Class I of the HDAC family comprises four members, HDAC-1, 2, 3, and 8. Class II of the HDAC family comprises HDAC-4, 5, 6, and 7, the molecular weights of which are all about two-fold larger than those of the class I members. Human HDAC-1, 2 and 3 were expressed in various tissues, but the others (HDAC-4, 5, 6, and 7) showed tissue-specific expression patterns. These results sμggest that each member of the HDAC family exhibits a different, individual substrate specificity and function in vivo.

Recommended Dilutions: Western Blot: 0.5-4 µg/ml

Type: Primary
Antigen: HDAC10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty